missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Displacement Vessel

Catalog No. S12929
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12929 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12929 Supplier Eisco™ Supplier No. PH0119A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For specific gravity experiment. The Glass Displacement Vessel features a spout.

  • Borosilicate glass vessel with spout
  • 500 mL capacity

Specifications

Grade 7 to Undergraduate
Includes 1 Displacement Vessel
Material Glass
Dimensions (L x W x H) 12.6 x 8.3 x 7.1 cm
Class Size 1 Lab Group
Product Type Displacement Vessel