Learn More
DWK Life Sciences Glass Ceramic Laboratory Protection Plate
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
- Due to low thermal expansion stresses, well suited to heating glassware with a Bunsen burner
- Protects cooking glasses from direct flame
Specifications
Specifications
| Material | Glass Ceramics |
| Autoclavable | Not Autoclavable |
| Dimensions (L x W) | 135 x 135 mm |
| For Use With (Application) | Heating glassware; Square Quadrupod for Glass Ceramic Laboratory Protection Plate |
| No. per Pack | 10/Pk. |