Learn More
DWK Life Sciences Glass Ceramic Laboratory Protection Plate
Catalog No. 09841026
Change view
Click to view available options
Dimensions (L x W):
135 x 135 mm
155 x 155 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
09841026 135 x 135 mm
09841027 155 x 155 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09841026 Supplier DWK Life Sciences Supplier No. 238215309
The glass ceramic plates are suitable for heating of cooking glasses with the Bunsen burner.
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • Due to low thermal expansion stresses, well suited to heating glassware with a Bunsen burner
  • Protects cooking glasses from direct flame
Specifications
Material Glass Ceramics
Autoclavable Not Autoclavable
Dimensions (L x W) 135 x 135 mm
For Use With (Application) Heating glassware; Square Quadrupod for Glass Ceramic Laboratory Protection Plate
No. per Pack 10/Pk.