missing translation for 'onlineSavingsMsg'
Learn More

Glass beads, 1.5 mm, Thermo Scientific Chemicals

$43.22 - $87.01

Specifications

Quantity 100 g
Chemical Name or Material Glass beads

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465941000
View Documents
Thermo Scientific Chemicals
465941000
100 g
Each for $43.22
Only null left
 
AC465945000
View Documents
Thermo Scientific Chemicals
465945000
500 g
Each for $87.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

100 g
Glass beads
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only