missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL45 Polypropylene Screw Caps

Catalog No. 064144
Change view
Click to view available options
Color:
Gray
Green
Light Gray
Purple
No. per Case:
10/Cs.
20/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
064144 Gray 10/Cs.
064143B Light Gray 20/Cs.
064143C Green 20/Cs.
064143A Purple 20/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 064144 Supplier Corning™ Supplier No. 139545LTMC
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Linerless plug seal design for efficient sealing

These are various sizes and types of screw caps for use with storage bottles.

Specifications

Color Gray
Includes Polytetrafluoroethylene (PTFE) Hydrophobic Membrane, 0.22μm. Allows sterile gas exchange while preventing contamination.
Material Polypropylene
No. per Case 10/Cs.
No. per Pack 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
For Use With (Equipment) Storage bottle
Autoclavable Autoclavable
Product Type Vented Screw Cap with Membrane