missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL45 Polypropylene Screw Caps

Catalog No. 064143B
Change view
Click to view available options
Color:
Gray
Green
Light Gray
Purple
No. per Case:
10/Cs.
20/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
064143B Light Gray 20/Cs.
064144 Gray 10/Cs.
064143C Green 20/Cs.
064143A Purple 20/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 064143B Supplier Corning™ Supplier No. 139545LTC2
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Linerless plug seal design for efficient sealing

These are various sizes and types of screw caps for use with storage bottles.

Specifications

Color Light Gray
Material Polypropylene
No. per Case 20/Cs.
No. per Pack 20/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
For Use With (Equipment) Storage bottle
Autoclavable Autoclavable
Product Type Plug Seal