Learn More
Corning™ GL45 Polypropylene Screw Caps
Linerless plug seal design for efficient sealing
Supplier: Corning™ 139545LTC1
Description
These are various sizes and types of screw caps for use with storage bottles.
Chemical Identifiers
| Polypropylene | |
| Non-sterile | |
| GL 45 | |
| Storage bottle |
Specifications
| Purple | |
| 20/Cs. | |
| Non-sterile | |
| Plug Seal | |
| Autoclavable |
| Polypropylene | |
| 20/Pk. | |
| GL 45 | |
| Storage bottle |