missing translation for 'onlineSavingsMsg'
Learn More

Girard's Reagent T 98.0+%, TCI America™

Catalog No. B0457500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0457500G 500 g
B045725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0457500G Supplier TCI America Supplier No. B0457500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-46-6
Molecular Formula C5H14ClN3O
Molecular Weight (g/mol) 167.64
MDL Number MFCD00012009
InChI Key YSULOORXQBDPCU-UHFFFAOYSA-N
Synonym girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide
PubChem CID 67156
IUPAC Name [(hydrazinecarbonyl)methyl]trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CC(=O)NN

Specifications

Melting Point 190°C
Color White
Quantity 500 g
Formula Weight 167.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Girard′s Reagent T
EINECSNumber (2)-3545
RTECSNumber CP3150000
TSCA Yes