missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Girard's Reagent D 99.0+%, TCI America™
Supplier: TCI America D072710G
Specifications
| Girard′s Reagent D | |
| 213°C | |
| C4H12N3O | |
| 10 g | |
| DALNXMAZDJRTPB-UHFFFAOYSA-O | |
| [(hydrazinecarbonyl)methyl]dimethylazanium | |
| 2733198 | |
| ≥99.0% (T) |
| 5787-71-3 | |
| White | |
| MFCD06656061 | |
| N,N-Dimethylglycine Hydrazide Dihydrochloride | |
| C[NH+](C)CC(=O)NN | |
| 118.16 | |
| 190.07 | |
| Crystalline Powder |
Chemical Identifiers
| 5787-71-3 | |
| 118.16 | |
| DALNXMAZDJRTPB-UHFFFAOYSA-O | |
| 2733198 | |
| C[NH+](C)CC(=O)NN |
| C4H12N3O | |
| MFCD06656061 | |
| N,N-Dimethylglycine Hydrazide Dihydrochloride | |
| [(hydrazinecarbonyl)methyl]dimethylazanium |
Safety and Handling
TSCA : Yes