Learn More
Ginkgolide B
Platelet-Activating Factor (PAF) antagonist | CAS: 15291-77-7 | C20H24O10 | 424.402 g/mol
Supplier: Thermo Scientific Chemicals J60646MF
Description
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGinkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.
Solubility
Soluble in DMSO. Slightly soluble in water and ethanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
Specifications
| Ginkgolide B | |
| White | |
| MFCD05662347 | |
| 14,4426 | |
| Soluble in DMSO; Slightly soluble in water and ethanol | |
| CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O | |
| 132470872 | |
| Powder |
| 15291-77-7 | |
| C20H24O10 | |
| 50 mg | |
| sqojoafxdqdrgf-maggbrlhsa-n | |
| SQOJOAFXDQDRGF-WVXYHUDCSA-N | |
| 424.402 | |
| 424.4 |
Chemical Identifiers
| 15291-77-7 | |
| 424.402 | |
| SQOJOAFXDQDRGF-WVXYHUDCSA-N | |
| 132470872 |
| C20H24O10 | |
| MFCD05662347 | |
| sqojoafxdqdrgf-maggbrlhsa-n | |
| CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only