Learn More
Gilson™ PIPETMANTM EcoPackTM Tips
Ensures optimal performance of PIPETMAN pipettes across the full volume range. GilsonTM PIPETMANTM EcoPackTM Tips are fully autoclavable.
Supplier:  Gilson™ F161670
 View more versions of this product
Catalog No. F161670G
Only null left
Explore available promotions
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk Packs are sealed with a tamper-proof strip which can easily be opened for quick access.
  • Bulk Packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EcoPack box contains 10,000 tips (10 bags of 1000 tips).
Specifications
Specifications
100 to 1000 μL
Non-sterile
Autoclavable
39.6 cm
Bagged
10000
Blue
Gilson Pipetman
Pipetman EcoPack
300 μL; 1000 μL
Non-filtered
Clear
15.59 in.
.85 mm
Universal
Pipet Tips
Single-use
Yes
Polypropylene
Product Suggestions
Product Suggestions