Learn More
Gilson™ EXPERTTM Tips, Tipack, Case
Universal fit EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. The tips are produced using state-of-the-art manufacturing techniques assuring lot-to-lot consistency and high performance.
$42.74 - $425.90
Specifications
| Filtered | Non-filtered |
|---|---|
| Clean Claims | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
| Format | Rack |
| Product Type | Air Displacement Tips |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Sterility | Volume (Metric) | Length (Metric) | Color | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Volume (Metric) | Length (Metric) | Color | Price | Quantity | ||
|
F1731002G
|
Gilson™
F1731002 |
Non-sterile | 0.1 to 10 μL | 2.88cm | Red |
Pack of 960 Each for $369.94
|
|
||
|
F1731012G
|
Gilson™
F1731012 |
Sterile | 0.1 to 10 μL | 2.88cm | Red |
Pack of 960 Each for $425.90
|
|
||
|
F1732012G
|
Gilson™
F1732012 |
Sterile | 0.1 to 10 μL | 4.53cm | Red |
Pack of 768 Each for $85.50
Case of 4 Pack for $342.00
|
|
||
|
F1732002G
|
Gilson™
F1732002 |
Non-sterile | 0.1 to 10 μL | 4.53cm | Red |
Pack of 768 Each for $73.84
Case of 4 Pack for $295.36
|
|
||
|
F1733012G
|
Gilson™
F1733012 |
Sterile | 1 to 200 μL | 5.14cm | Yellow |
Pack of 768 Each for $87.04
Case of 4 Pack for $348.16
|
|
||
|
F1733002G
|
Gilson™
F1733002 |
Non-sterile | 1 to 200 μL | 5.14cm | Yellow |
Case of 1000 Each for $295.34
|
|
||
|
F1735012G
|
Gilson™
F1735012 |
Sterile | 50 to 1250 μL | 10.22cm | Blue |
Pack of 384 Each for $42.74
Case of 8 Pack for $341.92
|
|
||
|
F1736002G
|
Gilson™
F1736002 |
Non-sterile | 1 to 300 μL | 5.85cm | Green |
Pack of 768 Each for $73.84
Case of 4 Pack for $295.36
|
|
||
|
F1736012G
|
Gilson™
F1736012 |
Sterile | 1 to 300 μL | 5.85cm | Green |
Pack of 768 Each for $85.50
Case of 4 Pack for $342.00
|
|
||
Description
Uniquely designed Tipacks feature a dual-purpose lid: hinged lid to easily open and close while using tips or pull pack out of hinges to set on top or discard for high-throughput tip usage.
- Fit leading pipetter brands
- Allow you to stock one brand of tip with practical packaging options so you can save time, money and bench space
- Produced in a sterile, clean-room environment using the purest raw materials and state-of-the-art manufacturing
- Offer consistent performance
- Free of Human DNA, RNase, DNase, PCR Inhibitors and pyrogens
- Feature an inert polymer hydrophobic surface to minimize sample binding when dispensing
- Clear packaging allows easy tip identification
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipetters
- Feature graduations at varying volumes allowing for quick checks when aspirating samples
- Economical and easy to use
Specifications
| Non-filtered | |
| Rack | |
| Single-use | |
| Yes | |
| Inert Polymer |
| DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free | |
| Air Displacement Tips | |
| Universal Fit | |
| EXPERT |