missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ EXPERTTM Standard Pipette Tips - Sterilized MegaPack
GREENER_CHOICE

Catalog No. F1733071G
Change view
Click to view available options
Graduations:
10, 50, 100μL
2, 5, 10μL
Length (Metric):
4.534cm
5.144cm
Model:
E200ST
EL10ST
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduations Length (Metric) Model
F1733071G 10, 50, 100μL 5.144cm E200ST
F1732071G 2, 5, 10μL 4.534cm EL10ST
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1733071G Supplier Gilson™ Supplier No. F1733071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Feature patented, low-retention polymer that creates hydrophobic surface to minimize sample binding while dispensing

  • For high-throughput multichannel pipetting
  • Features 192 tips per layer, 5 layers high
  • Gives choice to expose only 96 tips at a time w/portioned cover, or remove cover completely to access 192 tips
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens

Specifications

Description Sterilized MegaPack
Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model E200ST
Length (Metric) 5.144cm
Clean Claims RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
For Use With (Application) Pipetting
Format Rack
Tip Style Standard
Packaging Racked
Quantity 960/Cs.
Product Type Air Displacement Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line EXPERT
Material Polypropylene
Shape Conical
Graduations 10, 50, 100μL
Show More Show Less