missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN MTM Pipette Accessory, Charging Adapter

Catalog No. F807023G
Change view
Click to view available options
For Use With (Equipment):
Pipetteman™ M Carrousel™ pipette stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
F807023G Pipetteman™ M Carrousel™ pipette stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F807023G Supplier Gilson™ Supplier No. F807023

Choose this Gilson Charging Adapter when you need to provide continuous power to your PIPETMAN M while using a power carousel

Provides power during use or storage of pipette

Specifications

Color Dark Blue
For Use With Pipettes
Type Charging Adapter
For Use With (Equipment) Pipetteman™ M Carrousel™ pipette stand
Quantity 1
Width (English) 1.57 in