missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Accessory for PIPETMANTM Complete Pipetting System - Reagent Reservoir

Ideal for dispensing reagents with multichannel pipettes. GilsonTM Accessory for PIPETMANTM Complete Pipetting System- Reagent Reservoirs' unique shape allows for efficient and nearly complete sample withdrawal.

$509.88 - $509.88

Specifications

Color White
For Use With All Pipettes with Tips Mounted
Material Polystyrene
Type Reservoir
For Use With (Equipment) PIPETMAN™ Complete Pipetting System
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
F267670G
View Documents
Gilson™
F267670
50 mL
Pack of 10 Each for $509.88
Only null left
 
Description

Description

  • Angled sides and V-shaped bottoms
  • Pour-off spouts on all corners
  • Graduations on inside walls at middle volume
Specifications

Specifications

White
Polystyrene
PIPETMAN™ Complete Pipetting System
5.91 in
All Pipettes with Tips Mounted
Reservoir
100 Pk.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'