Learn More
Gilson™ Accessory for PIPETMANTM Complete Pipetting System - JimmyTM Microtube Openers
GREENER_CHOICE
Catalog No. F144983G
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
F144983G Polypropylene
1 options
Catalog No. F144983G Supplier Gilson™ Supplier No. F144983
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
JimmyTM Microtube Openers for use with PIPETMAN ClassicTM and PIPETMANTM Neo pipets.
  • For opening both snap-cap and screw-cap microtubes
  • Increases pipeting efficiency, lessens fatigue associated with manual tube opening and eliminates common mode of contamination
Specifications
For Use With All Pipettes with Tips Mounted
Material Polypropylene
Type Microtube Opener
For Use With (Equipment) PIPETMAN™ Complete Pipetting System
Quantity 3 Pk.