missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, FCC, 98-102%, Spectrum™ Chemical

Catalog No. 18604182
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604182 25 kg Poly Bottle
18604178 125 g Poly Bottle
18604179 500 g Poly Bottle
18604180 2.5 kg Poly Bottle
18604181 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604182 Supplier Spectrum Chemical Mfg Cor Supplier No. M110925KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1109, 13446-34-9, MnCl2·4H2O

Spectrum™ Chemical Manganese Chloride, Tetrahydrate, FCC is used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100%
Quantity 25 kg
Packaging Poly Bottle
Percent Purity 98 to 102%
Grade FCC
Chemical Name or Material Manganese Chloride, Tetrahydrate