missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain, Stock, for Staining of Blood, Tissue, and Malarial Parasites in Blood, Ricca Chemical

Catalog No. p-7239420
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32504 120 mL
325016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32504 Supplier Ricca Chemical Company Supplier No. RI32504
Only null left

Giemsa Stain for blood histopathology.

  • Components: Giemsa Stain, Glycerin, Methanol
  • Shelf life: 24 months

Used for blood staining and Malarial parasite staining applications

Specifications

Quantity 120 mL
Storage Requirements 15° to 30°C
Certifications/Compliance ISO, NIST
Format Liquid
Type Giemsa Stain
For Use With (Application) For Blood and tissue staining and staining of Malarial Parasites in Blood
Color Dark Violet
Packaging Natural Poly Bottle
CAS 56-81-5
Product Type Giemsa Stain