missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giemsa Stain, Stock, for Staining of Blood, Tissue, and Malarial Parasites in Blood, Ricca Chemical
Description
- Components: Giemsa Stain, Glycerin, Methanol
- Shelf life: 24 months
Used for blood staining and Malarial parasite staining applications
Specifications
Specifications
| Quantity | 120 mL |
| Storage Requirements | 15° to 30°C |
| Certifications/Compliance | ISO, NIST |
| Format | Liquid |
| Type | Giemsa Stain |
| For Use With (Application) | For Blood and tissue staining and staining of Malarial Parasites in Blood |
| Color | Dark Violet |
| Packaging | Natural Poly Bottle |
| CAS | 56-81-5 |
| Product Type | Giemsa Stain |