missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giemsa Stain Solution, Certified, LabChem™
Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | Giemsa Stain Solution |
| CAS | 56-81-5, 67-56-1, 51811-82-6 |
| Density | 1.03g/mL |
| CAS Max % | 60.98, 38.28, 0.74 |
| Packaging | Poly Bottle |
| Solubility | Soluble in water |
| Specific Gravity | 1.05g/mL ±0.02 |
| Decomposition Information | Hydrogen bromide, Bromine; Carbon monoxide; Carbon dioxide; Hydrogen chloride; Sulfur compounds |
| Color | Blue |
| Show More |