missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Giemsa Stain, Modified

Catalog No. 01288046
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288046 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288046 Supplier Sigma-Aldrich Supplier No. GS1284L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For hematology and histology purposes. Stains the nucleous and cytoplasm of white blood cells with blue or pink coloration.

When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.

  • Form: solution
  • Shelf life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Concentration: 0.4% (w/v) in buffered methanol solution
  • pH 6.8 with stabilizers
  • pH: 6.75 to 6.95
  • Applications: hematology and histology.
  • Storage temperature: room temperature

Specifications

Quantity 4 L
Product Type Giemsa Stain