missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Giemsa Stain, pure, high purity biological stain
CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol
$101.08 - $101.08
Chemical Identifiers
| CAS | 51811-82-6 |
|---|---|
| Molecular Formula | C14H14ClN3S |
| Molecular Weight (g/mol) | 291.80 |
| MDL Number | MFCD00012112,MFCD00081642 |
| InChI Key | NALREUIWICQLPS-UHFFFAOYSA-N |
| PubChem CID | 13735 |
| IUPAC Name | (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride |
| SMILES | [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC153230050
|
Thermo Scientific Chemicals
153230050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51811-82-6 | |
| 291.80 | |
| NALREUIWICQLPS-UHFFFAOYSA-N | |
| (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride |
| C14H14ClN3S | |
| MFCD00012112,MFCD00081642 | |
| 13735 | |
| [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1 |
Specifications
| 950 min. (in MeOH, on dried substance), 600 min. (in MeOH, on dried substance) A (1%/1cm) | |
| 51811-82-6 | |
| MFCD00012112,MFCD00081642 | |
| NALREUIWICQLPS-UHFFFAOYSA-N | |
| [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1 | |
| 291.80 | |
| 640 to 650nm (in MeOH), 520 to 525nm (in MeOH) | |
| Pure | |
| Black to Green | |
| Crystalline Powder or Crystals |
| 300.0°C | |
| C14H14ClN3S | |
| Solubility in water: soluble. Other solubilities: soluble in ethanol | |
| Authentic | |
| (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride | |
| 13735 | |
| 10% max. (110°C) | |
| Glass bottle | |
| 5 g | |
| Giemsa Stain |
Safety and Handling
EINECSNumber : 257-438-2
TSCA : TSCA
RUO – Research Use Only