missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Giemsa Stain, pure, high purity biological stain

Catalog No. AC153230250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153230250 25 g Glass bottle
AC153230050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153230250 Supplier Thermo Scientific Chemicals Supplier No. 153230250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Absorbance 950 min. (in MeOH, on dried substance), 600 min. (in MeOH, on dried substance) A (1%/1cm)
Melting Point 300.0°C
Infrared Spectrum Authentic
Lambda Max 640 to 650nm (in MeOH), 520 to 525nm (in MeOH)
Loss on Drying 10% max. (110°C)
Packaging Glass bottle
Color Black to Green
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Quantity 25 g
Physical Form Crystalline Powder or Crystals
Grade Pure
Chemical Name or Material Giemsa Stain
Show More Show Less
EINECSNumber 257-438-2
TSCA TSCA

RUO – Research Use Only