Learn More
Epredia™ Giemsa Stain
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
Supplier:  Epredia™ 89004
 View more versions of this product
Catalog No. 36-101-4353
Only null left
Description
Description
  • Erythrocytes stain pink with platelets exhibiting a very light, pale hue
  • Leukocyte nuclear chromatin stains magenta-purple
  • Monocyte cytoplasm stains the classical “ground glass” blue
  • Lymphocyte cytoplasm stains clear, sky blue
Specifications
Specifications
1 gal.
Giemsa Stain
Product Suggestions
Product Suggestions