missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giemsa's azur-eosin-methylene blue solution, For Microscopy, MilliporeSigma™
Description
For microscopy
Specifications
Specifications
| Chemical Name or Material | Giemsa's azur-eosin-methylene blue solution |
| Odor | Smells like methanol |
| UN Number | UN 1230 |
| Color | Blue |
| Quantity | 25 L |
| Physical Form | Liquid |