missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Distilled Water

Catalog No. p-4919257
Encompass
Change view
Click to view available options
Quantity:
5 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15230253 5 L
15230261 10 L
15230279 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15230253 Supplier Gibco™ Supplier No. 15230253
Only null left

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

  • Suitable for use in cell culture and molecular biology applications: This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Prepared by distillation and filtration (0.1μm), with no added substances
  • Water quality monitored on a routine basis: Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
  • Produced at a cGMP-compliant facility located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

pH 6 to 8
Grade Cell Culture
Packaging Universal Bag
Sterility Sterile-filtered
Treatment(s) Not DEPC-Treated
Quantity 5 L
Purification Method Membrane-Filtered
Shipping Condition Room Temperature
Product Line Gibco

For Research Use Only. Not for use in diagnostic procedures.