missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Schneider's Drosophila Medium
GREENER_CHOICE

Catalog No. 21720001
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21720001 10 x 500 mL
21720024 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21720001 Supplier Gibco™ Supplier No. 21720001
Only null left

Modified to contain L-glutamine and sodium bicarbonate

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells.

Formulation

  • Contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelos
  • Commonly supplemented with 10% fetal bovine serum (FBS) before use
  • Requires ambient CO2 in order to maintain physiological pH

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture

Compliance

Gibco Schneider's Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Specifications

Cell Line D.Mel-2
Cell Type Insect Cell
Form Liquid
Product Type Schneider's Drosophila Medium
Sterility Sterile
With Additives Glutamine
Without Additives No HEPES, No Phenol Red, No Sodium Pyruvate
Quantity 10 x 500 mL
Content And Storage Storage conditions: 2-8 °C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

For Research Use Only. Not for use in diagnostic procedures.