Learn More
Gibco™ RPMI 1640 Medium, HEPES

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
Supplier: Gibco™ 22400105
Requires : Supplementation
Description
RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This RPMI is modified as follows:
With: L-glutamine, HEPES, Phenol Red
The complete formulation is available.
Using RPMI
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Chemical Identifiers
| HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas | |
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Sterile-filtered | |
| No Sodium Pyruvate | |
| Gibco | |
| Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
| Leukemic Cells | |
| 1 X | |
| RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
| Sterile-filtered | |
| Glutamine, HEPES, Phenol Red | |
| cGMP-compliant under the ISO 13485 standard | |
| Room Temperature |
Specifications
| HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas | |
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Sterile-filtered | |
| No Sodium Pyruvate | |
| Gibco | |
| Room Temperature |
| Leukemic Cells | |
| 1 X | |
| RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
| Sterile-filtered | |
| Glutamine, HEPES, Phenol Red | |
| cGMP-compliant under the ISO 13485 standard | |
| 10 x 500 mL | |
| Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Safety and Handling
ShelfLife : 12 Months From Date of Manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.