missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement
GREENER_CHOICE

Catalog No. p-4919925
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61870036 500 mL
61870127 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61870036 Supplier Gibco™ Supplier No. 61870036
Only null left

Requires

Supplementation

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer.

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:

With: GlutaMAX™, Phenol Red

Without: HEPES

The complete formulation is available.

Using RPMI 1640 Medium

RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Cell Line HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell Type Leukemic Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives GlutaMAX, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco, GlutaMAX
Quantity 500 mL
Shipping Condition Room Temperature
Green Features ACT Ecolabel (Silver)
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.