Learn More
Gibco™ Opti-MEM™ Reduced Serum Medium, powder
Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology.
$518.00 - $532.00
Specifications
| Cell Line | Sp2, AE-1, CHO, BHK-21, and HEK |
|---|---|
| Cell Type | Primary Fibroblasts |
| Classification | Animal Origin |
| Form | Powder |
| Product Type | Opti-MEM I |
Description
Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications.
This Opti-MEM™ is modified as follows:
With: L-glutamine, Phenol Red
The complete formulation is confidential. For more information, please contact Technical Services.
Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%.
Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
| Sp2, AE-1, CHO, BHK-21, and HEK | |
| Animal Origin | |
| Opti-MEM I | |
| Glutamine, Phenol Red | |
| Gibco, Opti-MEM | |
| Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 30 months from date of manufacture |
| Primary Fibroblasts | |
| Powder | |
| Sterile-filtered | |
| cGMP-compliant under the ISO 13485 standard | |
| Room Temperature |
Safety and Handling
ShelfLife : 30 Months From Date of Manufacture
Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.