missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Neomycin Sulfate

Catalog No. 21810031
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21810031 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21810031 Supplier Gibco™ Supplier No. 21810031

Produced by Streptomyces fradiae

Polymyxin B Sulfate acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Gibco Neomycin Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria.

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 & Sf21 Cell Culture, Transformation

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Powder
Product Type Antibiotic
Sterility Non-sterile
For Use With (Application) Bacterial Selection
Quantity 100 g
Shipping Condition Room Temperature
Can Neomycin be used in mammalian selection?  Can Neomycin be used instead of Kanamycin in bacterial selection?

No, Neomycin is toxic to mammalian cells. It also causes irreversible damage to kidneys and other organs. Geneticin (aka G418 Sulfate) is a less toxic and very effective alternative for selection in mammalian cells.  Neomycin can be used in bacterial selection, but Kanamycin is the preferred drug to use because of Neomycin's toxicity.

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

For Research Use Only. Not for use in diagnostic procedures.