missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium NCTC-109 (1X)
GREENER_CHOICE

Catalog No. 21340039
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21340039 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21340039 Supplier Gibco™ Supplier No. 21340039

missing translation for 'validMainframeMsg'

Suitable for suspension and monolayer culture methods

NCTC-109 medium is a chemically defined medium that was originally developed by the National Cancer Institute and formulated for the culture of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco NCTC-109 medium is also suitable for generating and maintaining hybridoma cells. This NCTC-109 is modified as follows: With Phenol Red and Coenzymes. Without L-glutamine and HEPES. The complete formulation is available.

  • Gibco NCTC-109 medium is rich in nucleosides, vitamins, coenzymes, reducing agents and other unique ingredients
  • NCTC-109 contains no proteins, lipids, or growth factors. NCTC-109 medium typically requires supplementation of a growth factor cocktail (for serum free growth) or Fetal Bovine Serum
  • Uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

Specifications

Cell Type Hybridomas
Form Liquid
Product Type Cell Culture Medium
With Additives Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Pyruvate
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.