Learn More
Gibco™ McCoy's 5A (Modified) Medium

Description
Requires
Supplementation
McCoy's 5A (modified) Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.
This McCoy's 5A is modified as follows:
With: High Glucose, L-glutamine, Bacto-peptone, Phenol Red
Without: Sodium Pyruvate, HEPES
The complete formulation is available.
Using McCoy's 5A (modified) Medium
Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator. McCoy's 5A (modified) Medium requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). McCoy's 5A (modified) Medium uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Line | Rat fibroblasts |
| Cell Type | Biopsy Tissues |
| Classification | Animal Origin |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | McCoy's 5A Modified Medium |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | High Glucose, Glutamine, Phenol Red, Sodium Bicarbonate |
| Without Additives | No HEPES, No Sodium Pyruvate |
| Show More |
Frequently Asked Questions (FAQs)
We do not provide stability data for the product once it is opened as it would depend on the usage and storage conditions.
Yes, the medium should be stored in the dark because there are some components in the medium such as HEPES, Tryptophan, and Riboflavin, etc. that are sensitive to light.
Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.
We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.
Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.