missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ KaryoMAX™ Giemsa Stain Solution

Catalog No. 10092013
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10092013 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10092013 Supplier Gibco™ Supplier No. 10092013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For G-banding of chromosomes for cytogenetic analysis

  • Formulated as 6.0g Azur II Eosin and 1.6g Azur II per liter in glycerol/methanol
  • For in vitro diagnostic use
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Cell Culture, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Storage conditions: 15 – 30°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Form Liquid
Product Type Giemsa Stain
Sterility Sterile-filtered
For Use With (Application) Translational Research, Clinical Research
Product Line KaryoMAX
Quantity 100 mL
Shipping Condition Room Temperature

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.