missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Kanamycin Sulfate
GREENER_CHOICE

Catalog No. p-4918406
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 mL
Form:
Liquid
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
15160054 100 mL Liquid
11815024 5 g Powder
11815032 25 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15160054 Supplier Gibco™ Supplier No. 15160054
Only null left

Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus

  • A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
  • Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
  • Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
  • It is used for the prevention of bacterial contamination of cell cultures
  • Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
  • Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
  • Store at -5°C to -20°C

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Concentration 100 X, 10 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Bacterial Selection
Quantity 100 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.