missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM
GREENER_CHOICE

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells

$122.00 - $1,466.00

Specifications

Cell Line Jurkat, COS-7, and macrophage cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type IMDM (Iscove's Modified Dulbecco's Medium)
View More Specs

Requires: Supplementation


Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12440053
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
12440053
500 mL
Each for $161.00
Only null left
 
12440061
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
12440061
10 x 500 mL
Each for $1,280.00
Only null left
 
12440046
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
12440046
1000 mL
Each for $122.00
Only null left
 
12440079
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
12440079
6 x 1000 mL
Each for $1,466.00
Only null left
 
Description

Description

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: L-glutamine, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Jurkat, COS-7, and macrophage cells
1 X
IMDM (Iscove's Modified Dulbecco's Medium)
Sterile-filtered
No α-Thioglycerol, No 2-Mercaptoethanol
Gibco
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free
Liquid
Sterile-filtered
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months From Date of Manufacture

SDS
Documents

Documents

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.