missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-12 Nutrient Mix, powder

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells.

$126.00 - $425.00

Specifications

Cell Type Primary Rat Astrocytes
Form Powder
Product Type Ham's F-12 Nutrient Mixture
With Additives Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No HEPES, No Sodium Bicarbonate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
21700075
Encompass
View Documents Promotion Details
Gibco™
21700075
10 x 1 L
Each for $126.00
Only null left
 
21700018
Encompass
View Documents Promotion Details
Gibco™
21700018
50 L
Each for $425.00
Only null left
 
Description

Description

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Ham's F-12 is modified as follows:

With: L-glutamine, Phenol Red

Without: Sodium Bicarbonate

The complete formulation is available.

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations.

Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco F-12 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Specifications

Primary Rat Astrocytes
Ham's F-12 Nutrient Mixture
No HEPES, No Sodium Bicarbonate
Gibco
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Powder
Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 Months From Date of Manufacture

SDS
Documents

Documents

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.