missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bottle Weight

Catalog No. A1098801
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1098801 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1098801 Supplier Gibco™ Supplier No. A1098801
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keeps bottles upright in water baths to reduce risk of contamination

  • Fits 500mL and 1L Gibco bottle
  • Blue

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Color Blue
Content And Storage Storage conditions: 15 – 30°C
Shipping conditions: Ambient
For Use With (Equipment) 500mL and 1L Gibco Bottle
Product Line Gibco
Quantity 1 each
Shape Square
Shipping Condition Room Temperature
What is the internal diameter of the ring of your Bottle Weight (Cat. No. A1098801)?

The inner diameter is 2 1/16th inches and the entire width of the collar is 3 3/16th inches.

What material is the Bottle Weight (Cat. No. A1098801) made of?

The bottle weight is made of stainless steel with a plastic coating.

For Research Use Only. Not for use in diagnostic procedures.