missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ampicillin, sodium salt, irradiated
Description
Ampicillin differs from penicillin only by the presence of an amino group, which facilitates penetration through the outer membrane of some gram-negative bacteria. Ampicillin acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall.
- Used as selective antibiotic for resistant bacteria, generally at concentration of 10-25μg/ml for liquid media and 35-50μg/ml for plates
- Product is provided as powder
- Made into stock solution at 10mg/ml in water
- Ampicillin is not used to prevent bacterial contamination of cell cultures due to concerns of mammalian and insect cell toxicity
Cloning, Transformation
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility in Grand Island, NY. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Content And Storage | Storage conditions: 2 to 8°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
| Concentration | 20 to 125 μg/mL |
| Form | Powder |
| Product Type | Antibiotic |
| For Use With (Application) | Bacterial Selection |
| Quantity | 200 mg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.