missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Agarose (4%)
Description
- The 4% Agarose Gel melts at 65 to 70°C and remains fluid at 37°C
- When diluted with concentrated insect cell culture medium, Gibco™ 4% Agarose produces a gel firm enough to immobilize local infections, yet ductile enough to allow for the efficient removal of plaques for clonal expansion
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Storage conditions: 2 – 8°C. Protect from light. Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Concentration | 0.04 |
| Form | Gel |
| Product Type | Agarose |
| Sterility | Sterile |
| Sterilization Method | Autoclaved |
| For Use With (Application) | Plaque Assay |
| Quantity | 40 mL |
| Remarks | Not for electrophoresis |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.