missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced RPMI 1640 Medium
GREENER_CHOICE

Catalog No. p-4919817
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12633012 500 mL
12633020 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12633012 Supplier Gibco™ Supplier No. 12633012
Only null left

Requires

Supplementation

Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic RPMI 1640, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced RPMI 1640, with no adaptation, include Sp2, Vero, Raji, Daudi, and Jurkat.

This Advanced RPMI 1640 is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced RPMI 1640 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced RPMI 1640 requires supplementation with 1-5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced RPMI 1640 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iAdvanced Media: Reduced Sera Solutions. Quest, 2004. 1.3:26-30.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Advanced RPMI 1640 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Specifications

Cell Line Sp2, Vero, Raji, Daudi, and Jurkat
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level Reduced Serum
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.