missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced MEM
GREENER_CHOICE

Catalog No. p-4919816
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12492013 500 mL
12492021 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12492013 Supplier Gibco™ Supplier No. 12492013
Only null left

Requires

Supplementation

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic MEM, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced DMEM / F-12, with no adaptation, include PK-15, MDCK, Vero, WI-38 and Hep-2.

This Advanced MEM is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced MEM is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced MEM requires supplementation with 1-5% Fetal Bovine Serum and 4mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Advanced MEM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type MEM (Minimum Essential Medium)
Serum Level Reduced Serum
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Ambient
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.