Learn More
Giant Water Prism
Catalog No. S06667
Change view
Click to view available options
Includes:
Spout and instructions
Catalog No. Includes
S06667 Spout and instructions
1 options
Catalog No. S06667 Supplier Customer Channels Group (Ccg) Supplier No. 50300
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Spout and instructions
Fill transparent prism with water or mineral oil to produce a spectrum that the whole class can see.
  • Made from 0.25 in. thick acrylic
  • Height: 16 in.
  • Each panel is cut on a CNC machine and joined with the same techniques used for building aquariums
  • Guaranteed not to leak under normal use
Specifications
Product Type Water Prism
Includes Spout and instructions
For Use With (Application) Conduct colorful optics experiment
Subject Physics