missing translation for 'onlineSavingsMsg'
Learn More

Giant GeoSolids™

Catalog No. S79410
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79410 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79410 Supplier Customer Channels Group (Ccg) Supplier No. LER 3208

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Teacher's guide.

Make visual classroom demonstrations possible.

  • Fill with water, sand or rice as you explore shapes, sizes and the relationships between area and volume
  • Shapes include: cone, hemisphere, cube, cylinder, rectangular prism, hexagonal prism, triangular prism, square pyramid and triangular pyramid

Order Info

Grades 3+. Set of 10.

Specifications

For Use With (Application) For visual classroom demonstrations
Product Type Giant Geosolids
Grade 3+
Includes Cone, hemisphere, cube, cylinder, rectangular prism, hexagonal prism, triangular prism, square pyramid, triangular pyramid shapes, teacher's guide
Grade Level Elementary, Intermediate - Middle School, High School