missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ghrelin, Rat

Catalog No. AAJ64669MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64669MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64669MCR Supplier Thermo Scientific Chemicals Supplier No. J64669MCR

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 3314.78
Product Type Ghrelin, rat
Quantity 1 mg
Regulatory Status RUO - research use only

RUO – Research Use Only