missing translation for 'onlineSavingsMsg'
Learn More

Hardy Diagnostics™ QuickSlide™ HemaPRO Reagents Kit

Catalog No. 2300112
Change view
Click to view available options
Product Type:
Reagent Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
2300112 Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2300112 Supplier Hardy Diagnostics™ Supplier No. HP1BSK

Includes Wrights-Giemsa stain, buffer, and rinse for use with the Hardy Diagnostics™ QuickSlide™ HemaPRO automated hematology slide stainer

  • Includes Wrights-Giemsa stain, buffer, and rinse for use
  • Extended shelf life of 1 year (unopened)
  • Special formulated pack -125 tests/ Pk. in one self contained package

Recommended Storage

Wright Giemsa Stain - Keep container tightly closed. Store away from all sources of ignition. Store in a cool, dry, well-ventilated place

Specifications

Product Type Reagent Kit
Color Clear Bottles
Reagent Capacity 125 Tests, Stain Kit (3 Bottles)
For Use With (Application) QS-001 and QS-002
Includes Self-contained package with color-coded tubing
Weight (English) 5 lb.
Weight (Metric) 2.26 kg