Learn More
Abcam Inc anti-GFAP, Polyclonal, Abcam™ DFS Item
Supplier:  Abcam Inc AB726050UL
GFAP, Host Species: Rabbit, Target Species: Murine, Human, Mammalia, Feline, Porcine, Rat, Marmoset, Bovine, Polyclonal, Primary, Unconjugated, 50uL, Antisera, full length human recombinant GFAP expressed in bacteria and highly purified, IgG
Catalog No. NC9408158
Only null left
Explore available promotions