missing translation for 'onlineSavingsMsg'
Learn More

Gestodene 98.0+%, TCI America™

Catalog No. G0404100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0404100MG 100 mg
G04041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0404100MG Supplier TCI America Supplier No. G0404100MG

Chemical Identifiers

CAS 60282-87-3
Molecular Formula C21H26O2
Molecular Weight (g/mol) 310.44
MDL Number MFCD00867858
InChI Key SIGSPDASOTUPFS-KQMXEUTGSA-N
Synonym 13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one
PubChem CID 129317582
IUPAC Name (1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C

Specifications

Melting Point 199°C
Color White
Quantity 100 mg
Formula Weight 310.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gestodene
RTECSNumber JF7956000
TSCA No
Recommended Storage Freezer