Learn More
Bulbtronics™ Germicidal Lamp
Supplier:  Bulbtronics™ BBT194
Encompass
 View more versions of this product
Catalog No. 14395348
Only null left
Description
Description
Specifications
Chemical Identifiers
Germicidal UV-C
254 nm
Linear Double-ended Lamp
Specifications
Mini Bi-pin (G5)
7500 hr.
254 nm
1.58 cm
11.74 in.
T5
Germicidal UV-C
8 W
0.62 in.
Aqua fine 3010
29.84 cm
Linear Double-ended Lamp
Product Suggestions
Product Suggestions