Learn More
Germanium n-butoxide, 97+%
Oxide films | CAS: 25063-27-8
Supplier: Thermo Scientific Chemicals 04081606
Description
Germanium n-butoxide is used as oxide films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Germanium n-butoxide | |
| 1.017 g/mL | |
| 112°C (234°F) | |
| 1.4255 | |
| 5 g | |
| Tetra-n-butoxygermane | |
| 365.05 | |
| Liquid |
| 25063-27-8 | |
| 143°C (8 mmHg) | |
| Ge[O(CH2)3CH3]4 | |
| MFCD00015347 | |
| Moisture sensitive | |
| Hydrolyzes in water. | |
| ≥97% |
Chemical Identifiers
| 25063-27-8 | |
| MFCD00015347 | |
| Tetra-n-butoxygermane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 246-595-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only